The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138302.1 | NP_612175.1 | 3 - 4 | 443 | 49 |
GenBank mRNA | AB060274.1 | - | 3 - 4 | 443 | 49 |