Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slitrk4
Entrez Gene ID
245446
Gene Name
SLIT and NTRK-like family, member 4
Gene Aliases
6430513G13, D930039I09Rik
Chromosome Location
Chr.X: 64264883 - 64277367 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178740.4 NP_848855.2 2 - 3 260 83
GenBank mRNA AK039763.1 - 2 - 3 201 83
AK046301.1 - 2 - 3 130 83
AK053065.1 - 2 - 3 261 83
BC117890.1 - 2 - 3 125 83

View Full Product Details