The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033334.2 | NP_001028506.1 | 3 - 4 | 896 | 66 |
GenBank mRNA | AK140195.1 | - | 3 - 4 | 896 | 66 |
AY993933.1 | - | 4 - 5 | 945 | 66 | |
AY993934.1 | - | 3 - 4 | 920 | 66 | |
BC132221.1 | - | 2 - 3 | 613 | 66 | |
BC138501.1 | - | 2 - 3 | 613 | 66 |