Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbc1d24
Entrez Gene ID
224617
Gene Name
TBC1 domain family, member 24
Gene Aliases
9630033P11, C530046L02Rik, mKIAA1171
Chromosome Location
Chr.17: 24175431 - 24205562 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163847.1 NP_001157319.1 1 - 2 82 71
NM_001163848.1 NP_001157320.1 1 - 2 82 71
NM_001163849.1 NP_001157321.1 1 - 2 82 71
NM_001163850.1 NP_001157322.1 1 - 2 82 71
NM_001163851.1 NP_001157323.1 1 - 2 82 71
NM_173186.4 NP_775278.3 1 - 2 82 71
GenBank mRNA AK036088.1 - 1 - 2 35 71
AK045194.1 - 1 - 2 54 71
AK049754.1 - 1 - 2 56 71
AK083833.1 - 1 - 2 53 71
AK084693.1 - 1 - 2 36 71
AK086293.1 - 1 - 2 41 71
AK138431.1 - 1 - 2 43 71
AK147647.1 - 1 - 2 82 71

View Full Product Details