Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plxdc1
Entrez Gene ID
72324
Gene Name
plexin domain containing 1
Gene Aliases
2410003I07Rik, AI848450, Tem7
Chromosome Location
Chr.11: 97923237 - 97986667 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163608.1 NP_001157080.1 7 - 8 962 78
NM_028199.3 NP_082475.3 7 - 8 941 78
GenBank mRNA AK010361.1 - 7 - 8 941 78
AK036144.1 - 7 - 8 916 78
AK040620.1 - 6 - 7 818 78
AK153684.1 - 7 - 8 928 78

View Full Product Details