Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mmp25
Entrez Gene ID
240047
Gene Name
matrix metallopeptidase 25
Gene Aliases
F730048C11Rik, Leukolysin, MMP20, MT6-MMP
Chromosome Location
Chr.17: 23629458 - 23645269 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033339.3 NP_001028511.1 5 - 6 1416 75
GenBank mRNA AK089273.1 - 5 - 6 1155 75
AK089533.1 - 5 - 6 1155 75
AK137491.1 - 5 - 6 1018 75
AK154458.1 - 5 - 6 1418 75
AK171493.1 - 5 - 6 1155 75
BC059059.1 - 4 - 5 1486 75
BC112379.1 - 5 - 6 1102 75

View Full Product Details