The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_033185.1 | 1 - 2 | 401 | 127 | |
GenBank mRNA | AK013119.1 | - | 1 - 2 | 477 | 127 |
AK013540.1 | - | - | 181 | 127 | |
AK160881.1 | - | 1 - 2 | 123 | 127 | |
BB854398.1 | - | 1 - 2 | 402 | 127 |