The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029453.2 | NP_083729.2 | 7 - 8 | 1040 | 99 |
GenBank mRNA | AK133092.1 | - | 7 - 8 | 1041 | 99 |