Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sdccag8
Entrez Gene ID
76816
Gene Name
serologically defined colon cancer antigen 8
Gene Aliases
2700048G21Rik, 5730470G24Rik, Cccap
Chromosome Location
Chr.1: 176814526 - 177020437 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029756.3 NP_084032.1 16 - 17 2328 115
GenBank mRNA AK132316.1 - 16 - 17 2124 115
AK133622.1 - 16 - 17 2072 115

View Full Product Details