Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim69
Entrez Gene ID
70928
Gene Name
tripartite motif-containing 69
Gene Aliases
4921519C19Rik, Rnf36, Trif, Trimless
Chromosome Location
Chr.2: 122160700 - 122179027 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080510.2 NP_536771.2 3 - 4 770 88
GenBank mRNA AF334958.1 - 3 - 4 770 88
BC050815.1 - 3 - 4 762 88

View Full Product Details