The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003685.3 | NP_001003685.2 | 12 - 13 | 1236 | 65 |
GenBank mRNA | AK030504.1 | - | 12 - 13 | 1189 | 65 |
BC120748.1 | - | 12 - 13 | 1166 | 65 | |
BC120774.1 | - | 12 - 13 | 1166 | 65 | |
BC144892.1 | - | 12 - 13 | 1177 | 65 | |
L07379.1 | - | 12 - 13 | 1169 | 65 |