Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fastkd1
Entrez Gene ID
320720
Gene Name
FAST kinase domains 1
Gene Aliases
5330408N05Rik, mKIAA1800
Chromosome Location
Chr.2: 69686820 - 69712606 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177244.3 NP_796218.2 4 - 5 1072 96
GenBank mRNA AK030411.1 - - 1963 96
BC034292.1 - 1 - 2 179 96
BC069943.1 - 3 - 4 565 96
BC075689.1 - 4 - 5 1072 96

View Full Product Details