The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113574.1 | NP_001107046.1 | 9 - 10 | 1653 | 67 |
GenBank mRNA | AB206708.2 | - | 8 - 9 | 1464 | 67 |
AK037435.1 | - | 9 - 10 | 1635 | 67 | |
AK076472.1 | - | 9 - 10 | 1584 | 67 | |
AK076924.1 | - | 9 - 10 | 1733 | 67 |