Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kdm1b
Entrez Gene ID
218214
Gene Name
lysine (K)-specific demethylase 1B
Gene Aliases
4632428N09Rik, AI482520, Aof1
Chromosome Location
Chr.13: 47043471 - 47085279 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172262.3 NP_758466.1 15 - 16 1993 76
GenBank mRNA AK028553.1 - 5 - 6 659 76
AK078920.1 - 4 - 5 551 76
BC023917.1 - 15 - 16 1934 76

View Full Product Details