Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tshr
Entrez Gene ID
22095
Gene Name
thyroid stimulating hormone receptor
Gene Aliases
AI481368, hypothroid, hyt, pet
Chromosome Location
Chr.12: 91400993 - 91540509 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113404.1 NP_001106875.1 3 - 4 410 96
NM_011648.5 NP_035778.3 3 - 4 410 96
GenBank mRNA AK014519.1 - 3 - 4 409 96
AK021020.1 - 3 - 4 404 96
AK029084.1 - 3 - 4 376 96
AK041086.1 - 3 - 4 387 96
BB645263.1 - 3 - 4 410 96
BC028754.1 - 3 - 4 365 96
BC086691.1 - 3 - 4 339 96
BC092523.1 - 3 - 4 352 96
U02601.1 - 3 - 4 392 96
U02602.1 - 3 - 4 392 96

View Full Product Details