Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fkbp7
Entrez Gene ID
14231
Gene Name
FK506 binding protein 7
Gene Aliases
23kDa, FKBP-7, FKBP23
Chromosome Location
Chr.2: 76663034 - 76673139 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010222.2 NP_034352.1 3 - 4 576 147
GenBank mRNA AF040252.1 - 3 - 4 548 147
AK138924.1 - 3 - 4 492 147
BC032961.1 - 3 - 4 533 147

View Full Product Details