The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277296.1 | NP_001264225.1 | 10 - 11 | 1679 | 73 |
NM_001277297.1 | NP_001264226.1 | 10 - 11 | 1491 | 73 | |
NM_028105.4 | NP_082381.1 | 10 - 11 | 1600 | 73 | |
GenBank mRNA | AK011310.1 | - | 10 - 11 | 1451 | 73 |
AK050868.1 | - | 10 - 11 | 1581 | 73 | |
AK145987.1 | - | 7 - 8 | 1159 | 73 | |
AK154839.1 | - | 10 - 11 | 1546 | 73 | |
AK218268.1 | - | 2 - 3 | 249 | 73 | |
BC010539.1 | - | 10 - 11 | 1493 | 73 |