Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ak7
Entrez Gene ID
78801
Gene Name
adenylate kinase 7
Gene Aliases
4930502N02Rik, AK 7
Chromosome Location
Chr.12: 105703859 - 105782447 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030187.1 NP_084463.1 4 - 5 686 110
GenBank mRNA AK019664.1 - 4 - 5 551 110
AK029555.1 - 4 - 5 687 110

View Full Product Details