The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030187.1 | NP_084463.1 | 10 - 11 | 1276 | 65 |
GenBank mRNA | AK019664.1 | - | 10 - 11 | 1141 | 65 |
AK029555.1 | - | 10 - 11 | 1277 | 65 |