The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159317.1 | NP_001152789.1 | 4 - 5 | 725 | 73 |
NM_001159318.1 | NP_001152790.1 | 4 - 5 | 725 | 73 | |
NM_008364.2 | NP_032390.1 | 4 - 5 | 725 | 73 | |
NM_134103.2 | NP_598864.1 | 4 - 5 | 725 | 73 | |
GenBank mRNA | AK034269.1 | - | 4 - 5 | 689 | 73 |
AK039582.1 | - | 4 - 5 | 677 | 73 | |
AK045686.1 | - | 4 - 5 | 674 | 73 | |
AK083209.1 | - | 4 - 5 | 722 | 73 | |
AK136782.1 | - | 4 - 5 | 676 | 73 | |
AK149467.1 | - | 4 - 5 | 674 | 73 | |
BC021159.1 | - | 4 - 5 | 674 | 73 | |
X85999.1 | - | 4 - 5 | 676 | 73 |