Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mcm4
Entrez Gene ID
17217
Gene Name
minichromosome maintenance deficient 4 homolog (S. cerevisiae)
Gene Aliases
19G, AI325074, AU045576, Cdc21, Mcmd4, mKIAA4003, mcdc21
Chromosome Location
Chr.16: 15623897 - 15637400 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008565.3 NP_032591.3 14 - 15 2587 139
GenBank mRNA AK011743.1 - 14 - 15 2522 139
AK036653.1 - 14 - 15 2547 139
AK088796.1 - 14 - 15 2507 139
AK135422.1 - 14 - 15 2489 139
AK135619.1 - 2 - 3 287 139
AK141132.1 - 14 - 15 2555 139
AK141644.1 - 14 - 15 2588 139
AK146662.1 - 14 - 15 2510 139
AK151499.1 - 14 - 15 2501 139
AK157725.1 - 14 - 15 2489 139
AK220442.1 - 14 - 15 2489 139
BC013094.1 - 14 - 15 2543 139
D26089.1 - 14 - 15 2493 139

View Full Product Details