The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008565.3 | NP_032591.3 | 1 - 2 | 288 | 60 |
GenBank mRNA | AK011743.1 | - | 1 - 2 | 223 | 60 |
AK036653.1 | - | 1 - 2 | 247 | 60 | |
AK083618.1 | - | 1 - 2 | 202 | 60 | |
AK088796.1 | - | 1 - 2 | 208 | 60 | |
AK089999.1 | - | 1 - 2 | 192 | 60 | |
AK135422.1 | - | 1 - 2 | 190 | 60 | |
AK141132.1 | - | 1 - 2 | 254 | 60 | |
AK141644.1 | - | 1 - 2 | 289 | 60 | |
AK146662.1 | - | 1 - 2 | 211 | 60 | |
AK151499.1 | - | 1 - 2 | 202 | 60 | |
AK157725.1 | - | 1 - 2 | 190 | 60 | |
AK220442.1 | - | 1 - 2 | 189 | 60 | |
BC013094.1 | - | 1 - 2 | 244 | 60 | |
D26089.1 | - | 1 - 2 | 194 | 60 |