Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Etv5
Entrez Gene ID
104156
Gene Name
ets variant 5
Gene Aliases
1110005E01Rik, 8430401F14Rik, ERM
Chromosome Location
Chr.16: 22381313 - 22439570 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023794.2 NP_076283.2 6 - 7 497 72
GenBank mRNA AK018368.1 - 6 - 7 549 72
AK031452.1 - 6 - 7 544 72
AK049234.1 - 6 - 7 547 72
AK080674.1 - 6 - 7 474 72
AK164091.1 - 6 - 7 546 72
AK168881.1 - 6 - 7 545 72
AY004174.1 - 5 - 6 374 72
BC034680.1 - 6 - 7 497 72

View Full Product Details