Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Parn
Entrez Gene ID
74108
Gene Name
poly(A)-specific ribonuclease (deadenylation nuclease)
Gene Aliases
1200003I18Rik, DAN
Chromosome Location
Chr.16: 13537960 - 13668196 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028761.3 NP_083037.1 17 - 18 1308 78
GenBank mRNA AK004572.1 - 17 - 18 1309 78
AK041044.1 - 18 - 19 1368 78
AK045181.1 - 17 - 18 1294 78
AK160611.1 - 17 - 18 1277 78
BC021899.1 - 17 - 18 1276 78

View Full Product Details