Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Npffr2
Entrez Gene ID
104443
Gene Name
neuropeptide FF receptor 2
Gene Aliases
Gpr74, HG31, NPFF2
Chromosome Location
Chr.5: 89526577 - 89583914 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133192.3 NP_573455.2 2 - 3 439 66
GenBank mRNA AF236084.1 - 1 - 2 333 66
AF330054.1 - 1 - 2 324 66
AK050939.1 - 2 - 3 440 66

View Full Product Details