Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Grsf1
Entrez Gene ID
231413
Gene Name
G-rich RNA sequence binding factor 1
Gene Aliases
BB232551, C80280, D5Wsu31e
Chromosome Location
Chr.5: 88659233 - 88676171 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098476.2 NP_001091946.1 5 - 6 673 83
NM_178700.5 NP_848815.2 5 - 6 1001 83
GenBank mRNA AK044889.1 - 5 - 6 1002 83
AK045986.1 - 5 - 6 955 83
AK077782.1 - 5 - 6 1002 83
AK089014.1 - 4 - 5 537 83
AK168385.1 - 5 - 6 644 83
AK168459.1 - 5 - 6 644 83
AK210897.1 - 2 - 3 188 83
AK218546.1 - 2 - 3 188 83

View Full Product Details