The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139305.2 | NP_647466.2 | 10 - 11 | 1279 | 79 |
GenBank mRNA | AB086322.1 | - | 8 - 9 | 983 | 79 |
AJ245857.1 | - | 10 - 11 | 1281 | 79 | |
AK136579.1 | - | 10 - 11 | 1280 | 79 | |
AK137719.1 | - | 10 - 11 | 1281 | 79 | |
AK166403.1 | - | 7 - 8 | 708 | 79 | |
BC120544.1 | - | 10 - 11 | 1272 | 79 | |
BC120546.1 | - | 10 - 11 | 1272 | 79 | |
EF122497.1 | - | 8 - 9 | 987 | 79 |