Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem44
Entrez Gene ID
224090
Gene Name
transmembrane protein 44
Gene Aliases
1700007N03Rik, 9330161C17Rik, B230220N21
Chromosome Location
Chr.16: 30511855 - 30550834 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172614.3 NP_766202.3 9 - 10 1277 125
GenBank mRNA AK048655.1 - 9 - 10 1278 125
BC059216.1 - 9 - 10 1615 125

View Full Product Details