Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Afp
Entrez Gene ID
11576
Gene Name
alpha fetoprotein
Gene Aliases
-
Chromosome Location
Chr.5: 90490714 - 90508907 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007423.4 NP_031449.3 14 - 15 1890 83
GenBank mRNA AK010934.1 - 14 - 15 1866 83
AK075972.1 - 14 - 15 1869 83
AK076053.1 - 14 - 15 1866 83
AK076197.1 - 14 - 15 1868 83
AK145975.1 - 14 - 15 1866 83
AK146513.1 - 14 - 15 1868 83
AK146524.1 - 14 - 15 1868 83
AK167549.1 - 14 - 15 1866 83
AK168426.1 - 14 - 15 1869 83
AK168574.1 - 14 - 15 1868 83
AK168576.1 - 14 - 15 1866 83
AK168751.1 - 14 - 15 1868 83
AK168818.1 - 13 - 14 1744 83
AK168883.1 - 14 - 15 1866 83
AK169046.1 - 14 - 15 1868 83
AK169054.1 - 14 - 15 1868 83
AK169147.1 - 14 - 15 1866 83
AV105248.2 - 5 - 6 808 83
BC066206.1 - 14 - 15 1853 83
V00743.1 - 14 - 15 1864 83

View Full Product Details