Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmem8b
Entrez Gene ID
242409
Gene Name
transmembrane protein 8B
Gene Aliases
4930500O05Rik
Chromosome Location
Chr.4: 43668971 - 43692668 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085508.2 NP_001078977.1 4 - 5 729 63
GenBank mRNA AK029713.1 - 4 - 5 730 63
BC147569.1 - 4 - 5 600 63
BC147574.1 - 4 - 5 600 63
BC151070.1 - 4 - 5 600 63

View Full Product Details