The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028372.1 | NP_082648.1 | 1 - 2 | 568 | 58 |
GenBank mRNA | BU559276.1 | - | 1 - 2 | 565 | 58 |