The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134802.1 | NP_001128274.1 | 2 - 3 | 156 | 84 |
GenBank mRNA | AK134162.1 | - | 2 - 3 | 141 | 84 |