Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vmn1r60
Entrez Gene ID
636697
Gene Name
vomeronasal 1 receptor 60
Gene Aliases
EG636697, Gm7184
Chromosome Location
Chr.7: 5544197 - 5545099 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166732.1 NP_001160204.1 1 - 1 531 149

View Full Product Details