Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Eef1d
- Entrez Gene ID
-
66656
- Gene Name
- eukaryotic translation elongation factor 1 delta (guanine nucleotide exchange protein)
- Gene Aliases
-
EF-1-delta
- Chromosome Location
-
Chr.15: 75894796 - 75909556 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK006403.1 |
- |
1 - 2 |
27 |
61 |
View Full Product Details