The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010410.2 | NP_034540.1 | 2 - 2 | 529 | 83 |
GenBank mRNA | AF019566.1 | - | 2 - 2 | 530 | 83 |
AF041242.1 | - | 2 - 2 | 529 | 83 |