The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_212457.2 | NP_997622.1 | 3 - 3 | 478 | 68 |
GenBank mRNA | AK010400.1 | - | 3 - 3 | 481 | 68 |
AY833558.1 | - | 3 - 3 | 478 | 68 | |
BC090663.1 | - | 3 - 3 | 455 | 68 | |
BC116226.2 | - | 3 - 3 | 382 | 68 |