The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009987.4 | NP_034117.3 | 2 - 2 | 470 | 107 |
GenBank mRNA | AF102269.1 | - | 2 - 2 | 464 | 107 |
AK035902.1 | - | 2 - 2 | 445 | 107 | |
AK044557.1 | - | 2 - 2 | 471 | 107 | |
AK045634.1 | - | 2 - 2 | 458 | 107 | |
AK137388.1 | - | 2 - 2 | 458 | 107 | |
AK140010.1 | - | 2 - 2 | 444 | 107 | |
BC012653.1 | - | 2 - 2 | 426 | 107 |