The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024622.2 | NP_001019793.1 | 1 - 2 | 117 | 127 |
GenBank mRNA | AK145195.1 | - | 1 - 2 | 76 | 127 |
AK150595.1 | - | 1 - 2 | 78 | 127 | |
AK152279.1 | - | 1 - 2 | 80 | 127 | |
AK159201.1 | - | 1 - 2 | 88 | 127 | |
AK166071.1 | - | 1 - 2 | 101 | 127 | |
BC023320.1 | - | 1 - 2 | 88 | 127 | |
BC061235.1 | - | 1 - 2 | 81 | 127 | |
BC132588.1 | - | 1 - 2 | 139 | 127 | |
BC132590.1 | - | 1 - 2 | 139 | 127 | |
BY287179.1 | - | 1 - 2 | 117 | 127 |