Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Olfr62
Entrez Gene ID
18363
Gene Name
olfactory receptor 62
Gene Aliases
H12, IH12, MOR258-5
Chromosome Location
Chr.4: 118665519 - 118666466 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146315.2 NP_666427.2 1 - 1 29 103
GenBank mRNA BC104295.1 - 1 - 1 30 103

View Full Product Details