Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Abca16
Entrez Gene ID
233810
Gene Name
ATP-binding cassette, sub-family A (ABC1), member 16
Gene Aliases
AY243472
Chromosome Location
Chr.7: 120409647 - 120544892 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278944.1 NP_001265873.1 1 - 2 94 82
GenBank mRNA AY243472.1 - 1 - 2 94 82


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream ATP-binding cassette, sub-family A (ABC1), member 15 Abca15 4930500I12Rik -14.0kb Sense Chr. - 120328486 - 120407687 on Build GRCm38
Downstream Y box protein 1 pseudogene E130201H02Rik 173.7kb Sense Chr. - 120597625 - 120598475 on Build GRCm38

View Full Product Details