Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Synpo2
- Entrez Gene ID
-
118449
- Gene Name
- synaptopodin 2
- Gene Aliases
-
1110069I04Rik, 2310068J10Rik, 9530006G20Rik, AI848603, Myo
- Chromosome Location
-
Chr.3: 123076519 - 123236169 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080451.2 |
NP_536699.2 |
2 - 3 |
401 |
102 |
View Full Product Details