The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080451.2 | NP_536699.2 | 4 - 5 | 3377 | 76 |
GenBank mRNA | BC158045.1 | - | 4 - 5 | 3324 | 76 |