Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Rcsd1
- Entrez Gene ID
-
226594
- Gene Name
- RCSD domain containing 1
- Gene Aliases
-
A430105K13Rik, BC025872
- Chromosome Location
-
Chr.1: 165648945 - 165710063 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK030934.1 |
- |
1 - 1 |
2448 |
83 |
AK037254.1 |
- |
1 - 1 |
2454 |
83 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AI448597.1 |
- |
1 |
|
|
Chr. 1 - 165707066 - 165707440 [+] on Build GRCm38 |
83 |
GenBankmRNA |
DT926473.1 |
- |
1 |
325 |
|
Chr. 1 - 165706913 - 165707604 [-] on Build GRCm38 |
83 |
GenBankmRNA |
BX515655.1 |
- |
1 |
393 |
|
Chr. 1 - 165707154 - 165707671 [-] on Build GRCm38 |
83 |
GenBankmRNA |
BB151674.2 |
- |
1 |
470 |
|
Chr. 1 - 165707066 - 165707748 [-] on Build GRCm38 |
83 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
myelin protein zero-like 1 |
Mpzl1 |
1110007A10Rik, PZR |
-72.5kb
|
Sense |
Chr. - 165592181 - 165634558 on Build GRCm38 |
Downstream |
cellular repressor of E1A-stimulated genes 1 |
Creg1 |
AA755314, Creg |
54.0kb
|
Anti-sense |
Chr. - 165763732 - 165775304 on Build GRCm38 |
View Full Product Details