The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167592.2 | NP_001161064.1 | 1 - 2 | 133 | 65 |
GenBank mRNA | EU004071.1 | - | 1 - 2 | 133 | 65 |
EU703626.1 | - | 1 - 2 | 50 | 65 |