The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170884.1 | NP_001164355.1 | 7 - 7 | 1655 | 82 |
GenBank mRNA | C78977.1 | - | 1 - 1 | 82 |