The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039510.2 | NP_001034599.1 | 1 - 2 | 233 | 104 |
GenBank mRNA | AK038774.1 | - | 1 - 2 | 207 | 104 |
AK134532.1 | - | 1 - 2 | 236 | 104 |