Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gm4836
- Entrez Gene ID
-
22526
- Gene Name
- predicted gene 4836
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 28270710 - 28293632 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_009529.3 |
NP_033555.1 |
8 - 9 |
738 |
76 |
GenBank mRNA |
BC061169.1 |
- |
8 - 9 |
738 |
76 |
- Gene Symbol
- Gm10058
- Entrez Gene ID
-
100039240
- Gene Name
- predicted gene 10058
- Gene Aliases
-
ENSMUSG00000059047
- Chromosome Location
-
Chr.X: 27924021 - 27946885 on Build GRCm38
- Gene Symbol
- Gm10096
- Entrez Gene ID
-
100039377
- Gene Name
- predicted gene 10096
- Gene Aliases
-
ENSMUSG00000061227, Xmr-like
- Chromosome Location
-
Chr.X: 28931807 - 28954702 on Build GRCm38
- Gene Symbol
- Gm10147
- Entrez Gene ID
-
100039324
- Gene Name
- predicted gene 10147
- Gene Aliases
-
ENSMUSG00000064334
- Chromosome Location
-
Chr.X: 28584995 - 28607874 on Build GRCm38
- Gene Symbol
- Gm10230
- Entrez Gene ID
-
100042175
- Gene Name
- predicted gene 10230
- Gene Aliases
-
ENSMUSG00000068103
- Chromosome Location
-
Chr.X: 27472033 - 27494914 on Build GRCm38
- Gene Symbol
- Gm10486
- Entrez Gene ID
-
100039550
- Gene Name
- predicted gene 10486
- Gene Aliases
-
ENSMUSG00000073247
- Chromosome Location
-
Chr.X: 35067384 - 35090316 on Build GRCm38
- Gene Symbol
- Gm10487
- Entrez Gene ID
-
100039467
- Gene Name
- predicted gene 10487
- Gene Aliases
-
ENSMUSG00000073253
- Chromosome Location
-
Chr.X: 30820542 - 30843469 on Build GRCm38
- Gene Symbol
- Gm10488
- Entrez Gene ID
-
100042109
- Gene Name
- predicted gene 10488
- Gene Aliases
-
ENSMUSG00000073257
- Chromosome Location
-
Chr.X: 29976314 - 29999162 on Build GRCm38
- Gene Symbol
- Gm14525
- Entrez Gene ID
-
100039120
- Gene Name
- predicted gene 14525
- Gene Aliases
-
OTTMUSG00000017011
- Chromosome Location
-
Chr.X: 26672780 - 26702638 on Build GRCm38
- Gene Symbol
- Gm14632
- Entrez Gene ID
-
100042144
- Gene Name
- predicted gene 14632
- Gene Aliases
-
OTTMUSG00000017531
- Chromosome Location
-
Chr.X: 30329715 - 30352646 on Build GRCm38
- Gene Symbol
- Gm14819
- Entrez Gene ID
-
100039585
- Gene Name
- predicted gene 14819
- Gene Aliases
-
OTTMUSG00000018086
- Chromosome Location
-
Chr.X: 35404215 - 35427094 on Build GRCm38
- Gene Symbol
- Gm2012
- Entrez Gene ID
-
100039030
- Gene Name
- predicted gene 2012
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 26200882 - 26231323 on Build GRCm38
- Gene Symbol
- Gm2030
- Entrez Gene ID
-
100039065
- Gene Name
- predicted gene 2030
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 26287199 - 26310044 on Build GRCm38
- Gene Symbol
- Gm5168
- Entrez Gene ID
-
382275
- Gene Name
- predicted gene 5168
- Gene Aliases
-
EG382275
- Chromosome Location
-
Chr.X: 26028214 - 26051041 on Build GRCm38
- Gene Symbol
- Gm5169
- Entrez Gene ID
-
382277
- Gene Name
- predicted gene 5169
- Gene Aliases
-
Xmr
- Chromosome Location
-
Chr.X: 25277487 - 25301455 on Build GRCm38
- Gene Symbol
- Gm5935
- Entrez Gene ID
-
546282
- Gene Name
- predicted gene 5935
- Gene Aliases
-
EG546282
- Chromosome Location
-
Chr.X: 24753162 - 24775164 on Build GRCm38
- Gene Symbol
- Gm6121
- Entrez Gene ID
-
619991
- Gene Name
- predicted gene 6121
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 26912743 - 26935669 on Build GRCm38
- Gene Symbol
- Slx
- Entrez Gene ID
-
664829
- Gene Name
- Sycp3 like X-linked
- Gene Aliases
-
EG664958, Xmr
- Chromosome Location
-
Chr.X: 26522657 - 26545565 on Build GRCm38
View Full Product Details