Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Rhox3g
- Entrez Gene ID
-
546294
- Gene Name
- reproductive homeobox 3G
- Gene Aliases
-
OTTMUSG00000017157, Rhox3.7, Rhox3f, Rhox3g-ps
- Chromosome Location
-
Chr.X: 37623431 - 37628139 on Build GRCm38
- Gene Symbol
- Rhox3a
- Entrez Gene ID
-
382209
- Gene Name
- reproductive homeobox 3A
- Gene Aliases
-
AY147207, Gm1138, Prx, Rhox3, Rhox3.1
- Chromosome Location
-
Chr.X: 37249919 - 37258978 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_194063.3 |
NP_918952.2 |
5 - 6 |
813 |
77 |
GenBank mRNA |
DQ058640.1 |
- |
5 - 6 |
813 |
77 |
- Gene Symbol
- Rhox3c
- Entrez Gene ID
-
100135654
- Gene Name
- reproductive homeobox 3C
- Gene Aliases
-
EG665203, Rhox3.3, Rhox3b
- Chromosome Location
-
Chr.X: 37469868 - 37473961 on Build GRCm38
- Gene Symbol
- Rhox3e
- Entrez Gene ID
-
100135657
- Gene Name
- reproductive homeobox 3E
- Gene Aliases
-
ENSMUSG00000071772, OTTMUSG00000017148, Rhox3.5, Rhox3d
- Chromosome Location
-
Chr.X: 37546975 - 37550897 on Build GRCm38
- Gene Symbol
- Rhox3h
- Entrez Gene ID
-
434758
- Gene Name
- reproductive homeobox 3H
- Gene Aliases
-
EG434758, Rhox3.8, Rhox3g
- Chromosome Location
-
Chr.X: 37657688 - 37668764 on Build GRCm38
View Full Product Details