Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krtap4-2
Entrez Gene ID
68673
Gene Name
keratin associated protein 4-2
Gene Aliases
1110033F04Rik, AI604442
Chromosome Location
Chr.11: 99634114 - 99635084 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026807.2 NP_081083.2 1 - 1 54 90
GenBank mRNA AK004055.1 - 1 - 1 55 90

View Full Product Details