Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc48
Entrez Gene ID
74665
Gene Name
leucine rich repeat containing 48
Gene Aliases
4930449E07Rik
Chromosome Location
Chr.11: 60353206 - 60394342 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029044.2 NP_083320.1 12 - 13 1512 102
GenBank mRNA AK015430.1 - 12 - 13 1514 102
BC018412.1 - 12 - 13 1525 102

View Full Product Details